polyproLABS® Base Cabinets, Air Control
polyproLABS® Base Cabinets, Air Control
Supplier:   Air Control
polyproLABS® Base Cabinets, Air Control
Supplier:  Air Control
Certificates
polyproLABS® Base Cabinets, Air Control
polyproLABS® Base Cabinets, Air Control
85 Options Available Below
Product Details & Documents
Corrosion-resistant casework cabinets are constructed of stress-relieved, acid-resistant, 1.3 cm (½") thick polypropylene.
  • Drawers slide quietly and evenly on a polypropylene notched groove system
Drawer panels are 1 cm (⅜") thick and feature built-in stops to prevent accidental removal. The drawer suspension system is rated to withstand numerous cycles at 45.4 kg (100 lbs.). Doors are solid 1 cm (⅜") thick with beveled edges.
Two models of cabinets include adjustable shelves made from solid 1.3 cm (½") thick polypropylene. Cabinets with two half-width drawers and two doors include one shelf. Open-front bookcase units include two shelves. All other cabinets do not include shelves.
Joints are thermally welded by a special process that ensures leak-tight, flush unions. All hardware, including door hinges, screws, latches, drawer suspension systems, and handles, is constructed of polypropylene. No metals are used in cabinets. Toe space measures 10.2H×7.6D cm (4×3").
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Largeur
    Profondeur
    Hauteur
    Weight
    Availability
    Price
  • Base polyprolabs, 3 full-width drawers
    PP
    18"
    22"
    29"
    100 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,819.15
    $5,819.15
    Each
       $5,819.15
    Minimum order quantity is 1
    Supplier Number: AC-160-1830
  • Base polyprolabs, 3 full-width drawers
    PP
    24"
    22"
    29"
    110 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,974.35
    $5,974.35
    Each
       $5,974.35
    Minimum order quantity is 1
    Supplier Number: AC-160-2430
  • Base polyprolabs, 3 full-width drawers
    PP
    30"
    22"
    29"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,033.72
    $6,033.72
    Each
       $6,033.72
    Minimum order quantity is 1
    Supplier Number: AC-160-3030
  • Base polyprolabs, 3 full-width drawers
    PP
    36"
    22"
    29"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,093.04
    $6,093.04
    Each
       $6,093.04
    Minimum order quantity is 1
    Supplier Number: AC-160-3630
  • Base polyprolabs, 3 full-width drawers
    PP
    18"
    22"
    35"
    110 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,910.47
    $5,910.47
    Each
       $5,910.47
    Minimum order quantity is 1
    Supplier Number: AC-160-1836
  • Base polyprolabs, 3 full-width drawers
    PP
    24"
    22"
    35"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,033.72
    $6,033.72
    Each
       $6,033.72
    Minimum order quantity is 1
    Supplier Number: AC-160-2436
  • Base polyprolabs, 3 full-width drawers
    PP
    30"
    22"
    35"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,156.91
    $6,156.91
    Each
       $6,156.91
    Minimum order quantity is 1
    Supplier Number: AC-160-3036
  • Base polyprolabs, 3 full-width drawers
    PP
    36"
    22"
    35"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,280.15
    $6,280.15
    Each
       $6,280.15
    Minimum order quantity is 1
    Supplier Number: AC-160-3636
  • Base polyprolabs, 4 full-width drawers
    PP
    18"
    22"
    29"
    110 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,353.18
    $6,353.18
    Each
       $6,353.18
    Minimum order quantity is 1
    Supplier Number: AC-165-1830
  • Base polyprolabs, 4 full-width drawers
    PP
    24"
    22"
    29"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,476.38
    $6,476.38
    Each
       $6,476.38
    Minimum order quantity is 1
    Supplier Number: AC-165-2430
  • Base polyprolabs, 4 full-width drawers
    PP
    30"
    22"
    29"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,599.65
    $6,599.65
    Each
       $6,599.65
    Minimum order quantity is 1
    Supplier Number: AC-165-3030
  • Base polyprolabs, 4 full-width drawers
    PP
    36"
    22"
    29"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,718.29
    $6,718.29
    Each
       $6,718.29
    Minimum order quantity is 1
    Supplier Number: AC-165-3630
  • Base polyprolabs, 4 full-width drawers
    PP
    18"
    22"
    35"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,476.38
    $6,476.38
    Each
       $6,476.38
    Minimum order quantity is 1
    Supplier Number: AC-165-1836
  • Base polyprolabs, 4 full-width drawers
    PP
    24"
    22"
    35"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,599.65
    $6,599.65
    Each
       $6,599.65
    Minimum order quantity is 1
    Supplier Number: AC-165-2436
  • Base polyprolabs, 4 full-width drawers
    PP
    30"
    22"
    35"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,718.29
    $6,718.29
    Each
       $6,718.29
    Minimum order quantity is 1
    Supplier Number: AC-165-3036
  • Base polyprolabs, 4 full-width drawers
    PP
    36"
    22"
    35"
    150 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,841.51
    $6,841.51
    Each
       $6,841.51
    Minimum order quantity is 1
    Supplier Number: AC-165-3636
  • Base polyprolabs, 5 full-width drawers
    PP
    18"
    22"
    29"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,466.80
    $7,466.80
    Each
       $7,466.80
    Minimum order quantity is 1
    Supplier Number: AC-170-1830
  • Base polyprolabs, 5 full-width drawers
    PP
    24"
    22"
    29"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,589.93
    $7,589.93
    Each
       $7,589.93
    Minimum order quantity is 1
    Supplier Number: AC-170-2430
  • Base polyprolabs, 5 full-width drawers
    PP
    30"
    22"
    29"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,713.21
    $7,713.21
    Each
       $7,713.21
    Minimum order quantity is 1
    Supplier Number: AC-170-3030
  • Base polyprolabs, 5 full-width drawers
    PP
    36"
    22"
    29"
    150 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,836.48
    $7,836.48
    Each
       $7,836.48
    Minimum order quantity is 1
    Supplier Number: AC-170-3630
  • Base polyprolabs, 5 full-width drawers
    PP
    18"
    22"
    35"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,589.93
    $7,589.93
    Each
       $7,589.93
    Minimum order quantity is 1
    Supplier Number: AC-170-1836
  • Base polyprolabs, 5 full-width drawers
    PP
    24"
    22"
    35"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,713.21
    $7,713.21
    Each
       $7,713.21
    Minimum order quantity is 1
    Supplier Number: AC-170-2436
  • Base polyprolabs, 5 full-width drawers
    PP
    30"
    22"
    35"
    150 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,836.48
    $7,836.48
    Each
       $7,836.48
    Minimum order quantity is 1
    Supplier Number: AC-170-3036
  • Base polyprolabs, 5 full-width drawers
    PP
    36"
    22"
    35"
    160 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,955.18
    $7,955.18
    Each
       $7,955.18
    Minimum order quantity is 1
    Supplier Number: AC-170-3636
  • Base polyprolabs, 6 full-width drawers
    PP
    18"
    22"
    29"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,594.59
    $7,594.59
    Each
       $7,594.59
    Minimum order quantity is 1
    Supplier Number: AC-175-1830
  • Base polyprolabs, 6 full-width drawers
    PP
    24"
    22"
    29"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,717.80
    $7,717.80
    Each
       $7,717.80
    Minimum order quantity is 1
    Supplier Number: AC-175-2430
  • Base polyprolabs, 6 full-width drawers
    PP
    30"
    22"
    29"
    150 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,841.01
    $7,841.01
    Each
       $7,841.01
    Minimum order quantity is 1
    Supplier Number: AC-175-3030
  • Base polyprolabs, 6 full-width drawers
    PP
    36"
    22"
    29"
    160 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,959.75
    $7,959.75
    Each
       $7,959.75
    Minimum order quantity is 1
    Supplier Number: AC-175-3630
  • Base polyprolabs, 6 full-width drawers
    PP
    18"
    22"
    35"
    135 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,713.21
    $7,713.21
    Each
       $7,713.21
    Minimum order quantity is 1
    Supplier Number: AC-175-1836
  • Base polyprolabs, 6 full-width drawers
    PP
    24"
    22"
    35"
    145 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,841.01
    $7,841.01
    Each
       $7,841.01
    Minimum order quantity is 1
    Supplier Number: AC-175-2436
  • Base polyprolabs, 6 full-width drawers
    PP
    30"
    22"
    35"
    155 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,959.75
    $7,959.75
    Each
       $7,959.75
    Minimum order quantity is 1
    Supplier Number: AC-175-3036
  • Base polyprolabs, 6 full-width drawers
    PP
    36"
    22"
    35"
    165 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,082.93
    $8,082.93
    Each
       $8,082.93
    Minimum order quantity is 1
    Supplier Number: AC-175-3636
  • Base polyprolabs, 4 half-width, two full-width drawers
    PP
    36"
    22"
    29"
    160 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,265.55
    $8,265.55
    Each
       $8,265.55
    Minimum order quantity is 1
    Supplier Number: AC-180-3630
  • Base polyprolabs, 4 half-width, two full-width drawers
    PP
    42"
    22"
    29"
    170 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,388.77
    $8,388.77
    Each
       $8,388.77
    Minimum order quantity is 1
    Supplier Number: AC-180-4230
  • Base polyprolabs, 4 half-width, two full-width drawers
    PP
    48"
    22"
    29"
    180 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,511.92
    $8,511.92
    Each
       $8,511.92
    Minimum order quantity is 1
    Supplier Number: AC-180-4830
  • Base polyprolabs, 4 half-width, two full-width drawers
    PP
    36"
    22"
    35"
    165 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,635.18
    $8,635.18
    Each
       $8,635.18
    Minimum order quantity is 1
    Supplier Number: AC-180-3636
  • Base polyprolabs, 4 half-width, two full-width drawers
    PP
    42"
    22"
    35"
    175 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,753.80
    $8,753.80
    Each
       $8,753.80
    Minimum order quantity is 1
    Supplier Number: AC-180-4236
  • Base polyprolabs, 4 half-width, two full-width drawers
    PP
    48"
    22"
    35"
    185 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,877.10
    $8,877.10
    Each
       $8,877.10
    Minimum order quantity is 1
    Supplier Number: AC-180-4836
  • Base polyprolabs, 6 half-width, 1 full-width drawer
    PP
    36"
    22"
    29"
    160 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,393.24
    $8,393.24
    Each
       $8,393.24
    Minimum order quantity is 1
    Supplier Number: AC-185-3630
  • Base polyprolabs, 6 half-width, 1 full-width drawer
    PP
    42"
    22"
    29"
    170 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,699.00
    $8,699.00
    Each
       $8,699.00
    Minimum order quantity is 1
    Supplier Number: AC-185-4230
  • Base polyprolabs, 6 half-width, 1 full-width drawer
    PP
    48"
    22"
    29"
    180 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,004.88
    $9,004.88
    Each
       $9,004.88
    Minimum order quantity is 1
    Supplier Number: AC-185-4830
  • Base polyprolabs, 6 half-width, 1 full-width drawer
    PP
    36"
    22"
    35"
    165 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,758.41
    $8,758.41
    Each
       $8,758.41
    Minimum order quantity is 1
    Supplier Number: AC-185-3636
  • Base polyprolabs, 6 half-width, 1 full-width drawer
    PP
    42"
    22"
    35"
    175 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,064.17
    $9,064.17
    Each
       $9,064.17
    Minimum order quantity is 1
    Supplier Number: AC-185-4236
  • Base polyprolabs, 6 half-width, 1 full-width drawer
    PP
    48"
    22"
    35"
    185 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,369.97
    $9,369.97
    Each
       $9,369.97
    Minimum order quantity is 1
    Supplier Number: AC-185-4836
  • Base polyprolabs, 8 full-width drawers
    PP
    36"
    22"
    29"
    165 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,343.05
    $8,343.05
    Each
       $8,343.05
    Minimum order quantity is 1
    Supplier Number: AC-190-3630
  • Base polyprolabs, 8 full-width drawers
    PP
    42"
    22"
    29"
    175 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,767.54
    $8,767.54
    Each
       $8,767.54
    Minimum order quantity is 1
    Supplier Number: AC-190-4230
  • Base polyprolabs, 8 full-width drawers
    PP
    48"
    22"
    29"
    185 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,831.44
    $8,831.44
    Each
       $8,831.44
    Minimum order quantity is 1
    Supplier Number: AC-190-4830
  • Base polyprolabs, 8 full-width drawers
    PP
    36"
    22"
    35"
    195 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,831.44
    $8,831.44
    Each
       $8,831.44
    Minimum order quantity is 1
    Supplier Number: AC-190-3636
  • Base polyprolabs, 8 full-width drawers
    PP
    42"
    22"
    35"
    200 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,890.78
    $8,890.78
    Each
       $8,890.78
    Minimum order quantity is 1
    Supplier Number: AC-190-4236
  • Base polyprolabs, 8 full-width drawers
    PP
    48"
    22"
    35"
    205 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,954.63
    $8,954.63
    Each
       $8,954.63
    Minimum order quantity is 1
    Supplier Number: AC-190-4836
  • Base polyprolabs, 2 half-width/half-depth, 2 half-width, 2 full-width drawers
    PP
    36"
    22"
    29"
    175 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,831.44
    $8,831.44
    Each
       $8,831.44
    Minimum order quantity is 1
    Supplier Number: AC-195-3630
  • Base polyprolabs, 2 half-width/half-depth, 2 half-width, 2 full-width drawers
    PP
    42"
    22"
    29"
    180 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,137.19
    $9,137.19
    Each
       $9,137.19
    Minimum order quantity is 1
    Supplier Number: AC-195-4230
  • Base polyprolabs, 2 half-width/half-depth, 2 half-width, 2 full-width drawers
    PP
    48"
    22"
    29"
    185 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,442.99
    $9,442.99
    Each
       $9,442.99
    Minimum order quantity is 1
    Supplier Number: AC-195-4830
  • Base polyprolabs, 2 half-width/half-depth, 2 half-width, 2 full-width drawers
    PP
    36"
    22"
    35"
    200 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,196.51
    $9,196.51
    Each
       $9,196.51
    Minimum order quantity is 1
    Supplier Number: AC-195-3636
  • Base polyprolabs, 2 half-width/half-depth, 2 half-width, 2 full-width drawers
    PP
    42"
    22"
    35"
    205 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,502.36
    $9,502.36
    Each
       $9,502.36
    Minimum order quantity is 1
    Supplier Number: AC-195-4236
  • Base polyprolabs, 2 half-width/half-depth, 2 half-width, 2 full-width drawers
    PP
    48"
    22"
    35"
    210 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10,087.53
    $10,087.53
    Each
       $10,087.53
    Minimum order quantity is 1
    Supplier Number: AC-195-4836
  • Base polyprolabs, 1 full-width drawer, 1 door
    PP
    18"
    22"
    29"
    75 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,052.42
    $5,052.42
    Each
       $5,052.42
    Minimum order quantity is 1
    Supplier Number: AC-150-1830
  • Base polyprolabs, 1 full-width drawer, 1 door
    PP
    24"
    22"
    29"
    80 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,180.25
    $5,180.25
    Each
       $5,180.25
    Minimum order quantity is 1
    Supplier Number: AC-150-2430
  • Base polyprolabs, 1 full-width drawer, 1 door
    PP
    30"
    22"
    29"
    85 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,303.39
    $5,303.39
    Each
       $5,303.39
    Minimum order quantity is 1
    Supplier Number: AC-150-3030
  • Base polyprolabs, 1 full-width drawer, 1 door
    PP
    18"
    22"
    35"
    85 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,116.32
    $5,116.32
    Each
       $5,116.32
    Minimum order quantity is 1
    Supplier Number: AC-150-1836
  • Base polyprolabs, 1 full-width drawer, 1 door
    PP
    24"
    22"
    35"
    90 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,244.12
    $5,244.12
    Each
       $5,244.12
    Minimum order quantity is 1
    Supplier Number: AC-150-2436
  • Base polyprolabs, 1 full-width drawer, 1 door
    PP
    30"
    22"
    35"
    95 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,371.91
    $5,371.91
    Each
       $5,371.91
    Minimum order quantity is 1
    Supplier Number: AC-150-3036
  • Base polyprolabs, 2 half-width drawers, 2 doors
    PP
    24"
    22"
    35"
    100 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,942.41
    $5,942.41
    Each
       $5,942.41
    Minimum order quantity is 1
    Supplier Number: AC-155-2436
  • Base polyprolabs, 2 half-width drawers, 2 doors
    PP
    36"
    22"
    29"
    100 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,006.31
    $6,006.31
    Each
       $6,006.31
    Minimum order quantity is 1
    Supplier Number: AC-155-3630
  • Base polyprolabs, 2 half-width drawers, 2 doors
    PP
    42"
    22"
    29"
    110 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,083.91
    $6,083.91
    Each
       $6,083.91
    Minimum order quantity is 1
    Supplier Number: AC-155-4230
  • Base polyprolabs, 2 half-width drawers, 2 doors
    PP
    48"
    22"
    29"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,325.73
    $6,325.73
    Each
       $6,325.73
    Minimum order quantity is 1
    Supplier Number: AC-155-4830
  • Base polyprolabs, 2 half-width drawers, 2 doors
    PP
    36"
    22"
    35"
    110 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,102.13
    $6,102.13
    Each
       $6,102.13
    Minimum order quantity is 1
    Supplier Number: AC-155-3636
  • Base polyprolabs, 2 half-width drawers, 2 doors
    PP
    42"
    22"
    35"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,197.94
    $6,197.94
    Each
       $6,197.94
    Minimum order quantity is 1
    Supplier Number: AC-155-4236
  • Base polyprolabs, 2 half-width drawers, 2 doors
    PP
    48"
    22"
    35"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,453.61
    $6,453.61
    Each
       $6,453.61
    Minimum order quantity is 1
    Supplier Number: AC-155-4836
  • Open-front bookcase, without doors
    PP
    30"
    13"
    29"
    50 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,290.26
    $4,290.26
    Each
       $4,290.26
    Minimum order quantity is 1
    Supplier Number: AC-120-3030
  • Open-front bookcase, without doors
    PP
    36"
    13"
    29"
    55 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,477.39
    $4,477.39
    Each
       $4,477.39
    Minimum order quantity is 1
    Supplier Number: AC-120-3630
  • Open-front bookcase, without doors
    PP
    42"
    13"
    29"
    60 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,669.07
    $4,669.07
    Each
       $4,669.07
    Minimum order quantity is 1
    Supplier Number: AC-120-4230
  • Open-front bookcase, without doors
    PP
    48"
    13"
    29"
    65 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,865.32
    $4,865.32
    Each
       $4,865.32
    Minimum order quantity is 1
    Supplier Number: AC-120-4830
  • Open-front bookcase, without doors
    PP
    30"
    13"
    35"
    55 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,349.53
    $4,349.53
    Each
       $4,349.53
    Minimum order quantity is 1
    Supplier Number: AC-120-3036
  • Open-front bookcase, without doors
    PP
    36"
    13"
    35"
    65 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,545.80
    $4,545.80
    Each
       $4,545.80
    Minimum order quantity is 1
    Supplier Number: AC-120-3636
  • Open-front bookcase, without doors
    PP
    42"
    13"
    35"
    65 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,732.92
    $4,732.92
    Each
       $4,732.92
    Minimum order quantity is 1
    Supplier Number: AC-120-4236
  • Open-front bookcase, without doors
    PP
    48"
    13"
    35"
    70 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,865.32
    $4,865.32
    Each
       $4,865.32
    Minimum order quantity is 1
    Supplier Number: AC-120-4836
  • Base cabinets, 2 doors, for blind corners
    PP
    53"
    22"
    29"
    135 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,650.33
    $5,650.33
    Each
       $5,650.33
    Minimum order quantity is 1
    Supplier Number: AC-115-3030
  • Base cabinets, 2 doors, for blind corners
    PP
    59"
    22"
    29"
    170 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,819.15
    $5,819.15
    Each
       $5,819.15
    Minimum order quantity is 1
    Supplier Number: AC-115-3630
  • Base cabinets, 2 doors, for blind corners
    PP
    65"
    22"
    29"
    150 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,070.25
    $6,070.25
    Each
       $6,070.25
    Minimum order quantity is 1
    Supplier Number: AC-115-4230
  • Base cabinets, 2 doors, for blind corners
    PP
    71"
    22"
    30"
    160 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,070.25
    $6,070.25
    Each
       $6,070.25
    Minimum order quantity is 1
    Supplier Number: AC-115-4830
  • Base cabinets, 2 doors, for blind corners
    PP
    53"
    22"
    35"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,942.41
    $5,942.41
    Each
       $5,942.41
    Minimum order quantity is 1
    Supplier Number: AC-115-3036
  • Base cabinets, 2 doors, for blind corners
    PP
    59"
    22"
    35"
    155 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,942.41
    $5,942.41
    Each
       $5,942.41
    Minimum order quantity is 1
    Supplier Number: AC-115-3636
  • Base cabinets, 2 doors, for blind corners
    PP
    65"
    22"
    35"
    135 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,197.94
    $6,197.94
    Each
       $6,197.94
    Minimum order quantity is 1
    Supplier Number: AC-115-4236
  • Base cabinets, 2 doors, for blind corners
    PP
    71"
    22"
    35"
    145 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,197.94
    $6,197.94
    Each
       $6,197.94
    Minimum order quantity is 1
    Supplier Number: AC-115-4836
Accessories
Informations sur le produit
  • Catalog #
    Material
    Height
    Availability
    Price
  • Pegboard Drying Rack
    Pegboard Drying Rack
    89008-208
    PP
    6"
    Log in pour la disponibilité
    Each - $1,363.59
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Front filler panel
    Front filler panel
    89008-778
    Log in pour la disponibilité
    Each - $448.87
Informations sur le produit
  • Catalog #
    Availability
    Price
  • End filler panel
    End filler panel
    89008-780
    Log in pour la disponibilité
    Each - $406.23
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Countertop
    Countertop
    89008-782
    Log in pour la disponibilité
    Each - $512.12