ThermoSafe® Absorbent Sheets, ThermoSafe Brands
ThermoSafe® Absorbent Sheets, ThermoSafe Brands
Supplier:   Sonoco Thermosafe
ThermoSafe® Absorbent Sheets, ThermoSafe Brands
Supplier:  Sonoco Thermosafe
Certificates
ThermoSafe® Absorbent Sheets, ThermoSafe Brands
ThermoSafe® Absorbent Sheets, ThermoSafe Brands
3 Options Available Below
Product Details & Documents
Sheets absorb spills that may occur during shipment of diagnostic specimens and biological materials.
  • Spill control material
  • Meets packaging and transportation regulations
Made of tissue-based paper substrate impregnated with super-absorbent polymers to help prevent leakage and possible contamination of package handler. Sheets meet regulations and recommendations of U.S. Government, U.S. Postal Service, and United Nations for goods packaging and transporation of diagnostic specimens, biological materials, and other dangerous goods.
Product Family Options
Product Information
  • Capacité
    Details
    Dimensions
    Availability
    Price
  • 22.5 ml (0.8 oz.)
    Water Capacity 90 mL (3 oz.)
    7.6L x 7.6W cm (3 x 3")
    Log in for availability
    Log in for availability
    Case of 10,000
    Case of 10,000 - $996.70
    $996.70
    New Customer Promotional Price
    Case of 10,000
       $996.70
    Supplier Number: ZORB33
  • 90 ml (3 oz.)
    Water Capacity 360 mL (12.2 oz.)
    15.2L x 15.2W cm (6 x 6")
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $282.25
    $282.25
    New Customer Promotional Price
    Case of 500
       $282.25
    Supplier Number: ZORB66
  • 360 ml (12.2 oz.)
    Water Capacity 1440 mL (48.7 oz.)
    30.5L x 30.5W cm (12 x 12")
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $936.00
    $936.00
    New Customer Promotional Price
    Case of 500
       $936.00
    Supplier Number: ZORB1212