InLab® Routine Pro 3-in-1 pH Electrodes, METTLER TOLEDO®
General-purpose electrodes feature integrated temperature probe and ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures
- ARGENTHAL™ lead-off and silver ion trap
- High alkali glass body
- Resistant to corrosives
High alkali glass body is resistant to organic or corrosive samples. Supplied with 25mL electrolyte vial. Diameter: 12mm.
Frequently Bought Together