Cushion-Grade Static-Dissipative Foam, Protektive Pak®
Fournisseur: Protektive Pak
Certificats
À propos de cet article
Static-dissipative crosslink polyethylene foam protects items from physical shock.
- Dissipative material is the preferred packaging for ESD-sensitive items in intimate contact
- Features a static-dissipative surface of 107–109 ohms/sq., which is ideal for controlled electrostatic charge removal
- Foam is non-contaminating, non-corrosive, and non-sloughing
- Easy to adapt for custom uses by die-cutting, laminating, routing, and more
Density: 30.4 kg/m3 (1.9 lbs./cu.ft.)
Frequently Bought Together