BioCoat™ HTS Caco-2 Assay Systems, PET Membrane, Corning
Catalog # CA-CB354803
Supplier: Corning
Specifications
- Description:HTS Caco-2 assay system, inc. media and supplements, for 120 individual 3-day Caco-2 assays
- Cat. no.:CA-CB354803
- Supplier No.:354802
Specifications
About this item
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 assay system, PET membrane contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat fibrillar collagen 24-well insert system.