Microslides Electrodes, BTX® | Avantor
Microslides Electrodes, BTX®
Supplier:   Harvard Apparatus
Microslides Electrodes, BTX®
Supplier:  Harvard Apparatus
Certificates
Microslides Electrodes, BTX®
Microslides Electrodes, BTX®
4 Options Available Below
Product Details & Documents
Ideal for embryo manipulation, nuclear transfer, hybridoma production, and cell or protoplast fusion
Stainless steel electrodes are mounted on glass slides designed to easily fit on a microscope to allow observation of the dimer formation during electrofusion.
Ordering information: Connection to an electroporator requires banana plug-to-banana plug connector cable (82019-194).
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Gap Size
    Availability
    Price
  • Glass Microslides with rectangular electrodes
    2.0 ml
    10 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $575.44
    $575.44
    Each
       $575.44
    Supplier Number: 45-0106
  • Divergent field
    20 µl
    0.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $3,355.56
    $3,355.56
    Pack of 10
       $3,355.56
    Supplier Number: 45-0103
  • Divergent field
    40 µl
    1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $3,405.88
    $3,405.88
    Pack of 10
       $3,405.88
    Supplier Number: 45-0104
  • Homogenous field
    650 µl
    3.2 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $818.04
    $818.04
    Each
       $818.04
    Supplier Number: 45-0105
Accessories
Informations sur le produit
  • Catalog #
    Color
    Length
    Availability
    Price
  • Minigrabber® ECM 2001 to banana cables
    Minigrabber® ECM 2001 to banana cables
    82019-194
    Black/red
    3 m (10 ft.)
    Log in pour la disponibilité
    Each - $343.38