2-Needle Array™ Electrode Kits, BTX®
Supplier:   Harvard Apparatus
2-Needle Array™ Electrode Kits, BTX®
Supplier:  Harvard Apparatus
Certificates
2-Needle Array™ Electrode Kits, BTX®
2 Options Available Below
Product Details & Documents
In vivo-style electrodes are designed specifically for muscle gene delivery
Kit 22221-522 is recommended for larger muscle masses, such as rat gastrocnemus. Kit 22221-524 is recommended for smaller muscle masses, such as mouse tibialis.
Electrodes may be used with electroporators 58018-004, 58017-932, and 58017-984. The ECM®/Enhancer series (82019-182) is recommended for monitoring the electrodes.
Electrode consists of a reusable Delrin® plastic handle and a sterile, disposable two-needle array assembly. Needle array assemblies are packaged in “six packs,” allowing for guaranteed sterility. Needles are made of medical-grade stainless steel.
 : Kit 22221-522 includes a 10mm diameter two-needle array handle and a sterile package of six electrode assemblies with 10mm spacing. Kit 22221-524 includes a 5mm diameter handle and a sterile package of six electrode assemblies with 5mm spacing.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 22221-522
  • 22221-524
  • Voltage range:
  • 0 - 500 V
  • 0 - 500 V
  • Pulse length range:
  • 1 µs to 99 ms in PBS (Phosphate Buffer Saline)
  • 1 µs to 99 ms in PBS (Phosphate Buffer Saline)
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Handle Length
    Needle Length
    Availability
    Price
  • 2-Needle Array™, electrode gap 10 mm
    8 cm
    20 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,122.24
    $1,122.24
    Each
       $1,122.24
    Supplier Number: 45-0167
    Spécifications:
    More
    Specifications
    Cat. No.
    22221-522
    Voltage range
    0 - 500 V
    Pulse length range
    1 µs to 99 ms in PBS (Phosphate Buffer Saline)
  • 2-Needle Array™, electrode gap 5 mm
    8 cm
    20 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,059.51
    $1,059.51
    Each
       $1,059.51
    Supplier Number: 45-0168
    Spécifications:
    More
    Specifications
    Cat. No.
    22221-524
    Voltage range
    0 - 500 V
    Pulse length range
    1 µs to 99 ms in PBS (Phosphate Buffer Saline)