Caliper Electrodes, BTX
À propos de cet article
In vivo-style, reusable electrodes are suitable for a variety of applications, including gene and transdermal drug delivery
Electrodes are compatible with electroporation systems 58018-004 and 58017-932. Electrodes complement Tweezertrodes™ 22221-518 and -520; Genetrodes® 47727-356 and -358; and 2-Needle Array™ electrode kits 22221-522 and -524.
Calipers may be cleaned with a mild detergent and sterilized with ethanol or ethylene oxide.
Each model consists of a caliper and a pair of end plates. End plates are adjusted using the black roller mounted on the caliper. Use brass caliper for smaller animals with a smaller surface area. Use stainless steel calipers for slightly larger surface areas.
2 Options Available Below
Product Details & Documents
In vivo-style, reusable electrodes are suitable for a variety of applications, including gene and transdermal drug delivery
Electrodes are compatible with electroporation systems 58018-004 and 58017-932. Electrodes complement Tweezertrodes™ 22221-518 and -520; Genetrodes® 47727-356 and -358; and 2-Needle Array™ electrode kits 22221-522 and -524.
Calipers may be cleaned with a mild detergent and sterilized with ethanol or ethylene oxide.
Each model consists of a caliper and a pair of end plates. End plates are adjusted using the black roller mounted on the caliper. Use brass caliper for smaller animals with a smaller surface area. Use stainless steel calipers for slightly larger surface areas.
Specifications for Product Family
Spécifications
- Catalog No:
- 47727-350
- 47727-352
- Voltage range:
- 0 - 500 V
- 0 - 500 V
- Pulse length range:
- 1µs–99ms
- 1µs–99ms
Spécifications
Product Family Options
Informations sur le produit
- DésignationDimensionsGap SizeAvailabilityPrice
- Supplier Number: 45-0102Spécifications:
More
Specifications
Cat. No.47727-350Voltage range0 - 500 VPulse length range1µs–99ms - Supplier Number: 45-0101Spécifications:
More
Specifications
Cat. No.47727-352Voltage range0 - 500 VPulse length range1µs–99ms