Petri Dish Electrode, BTX®
Petri Dish Electrode
Catalog # 58017-970
Supplier: Harvard Apparatus
Fournisseur #: 45-0100
Jump to:
Product Details & Documents
For use with a 100mm petri dish that functions as the electroporation chamber
Can be used for electroporation of tissues grown in the petri dish. Compatible with BTX ECM® 630 and 830 systems. Electrodes may be rinsed with mild detergent and deionized water, and sterilized with 70% ethanol.
Electrodes are designed to electroporate volumes ranging from 4mL to 60mL. Electrode assemblies contain parallel stainless steel electrodes, which produce homogeneous fields.
Spécifications
- Désignation:Petri Dish Electrode