LE703 Four-Pole Graphite Conductivity Probe, METTLER TOLEDO®
Catalogue # CA97002-308
Fournisseur: Mettler Toledo
Spécifications
- Ø:12 mm
- Temperature Range:0...80 °C
- Description:Electrode
- Length:120 mm
- Cat. no.:CA97002-308
- Body Material:Plastic
- Range:10 µS/cm - 500 mS/cm
Spécifications
À propos de cet article
General-purpose probe is designed for use with METTLER TOLEDO® conductivity meters in a laboratory setting.
- For conductivity testing
- Four graphite poles
- Plastic shaft
Probe features four graphite poles embedded into plastic shaft to eliminate polarization.