Conductivity Cells, WTW
About this item
This selection of conductivity cells includes the TetraCon® series of cells, which feature the highest degree of precision and linearity by optimized cell geometry, an extremely large measuring range with just one cell, long-term cell constant stability with high-quality abrasion-resistant graphite electrodes, and a built-in temperature sensor
The flow-thru vessel is constructed of transparent PVC and has a 18mm I.D. Filling volume (without sensor): 13mL.
TetraCon® cells also feature the smallest immersion depth possible and have no measuring errors due to cable influences, primary or secondary polarization effects, or contact with sidewalls or base of measuring vessels.
7 Options Available Below
Product Details & Documents
This selection of conductivity cells includes the TetraCon® series of cells, which feature the highest degree of precision and linearity by optimized cell geometry, an extremely large measuring range with just one cell, long-term cell constant stability with high-quality abrasion-resistant graphite electrodes, and a built-in temperature sensor
The flow-thru vessel is constructed of transparent PVC and has a 18mm I.D. Filling volume (without sensor): 13mL.
TetraCon® cells also feature the smallest immersion depth possible and have no measuring errors due to cable influences, primary or secondary polarization effects, or contact with sidewalls or base of measuring vessels.
Product Family Options
Product Information
- DésignationMatièreØCable LengthBody MaterialCell ConstantAvailabilityPrice
- Supplier Number: 203730
- TetraCon 325/SGraphite15.3 mm (5/8")1.5 m (5 ft.)Epoxy0.491 cm⁻¹Log in for availabilityEach$3,052.71Supplier Number: 301602
- Ultrapure Water Cell LR 325/01V4A Steel12 mm (1/2")1.5 m (5 ft.)V4A Steel0.1 cm⁻¹Log in for availabilityEach$2,656.98Supplier Number: 301961
- Trace Cell LR 325/001V4A Steel20 mm (13/16")1.5 m (5 ft.)V4A Steel0.01 cm⁻¹Log in for availabilityEach$4,521.50Supplier Number: 301962
- TetraCon 325-3Graphite15.3 mm (5/8")3 m (10 ft.)Epoxy0.475 cm⁻¹Log in for availabilityEach$2,367.49Supplier Number: 301970
- TetraCon 325-6Graphite15.3 mm (5/8")6 m (20 ft.)Epoxy0.475 cm⁻¹Log in for availabilityEach$2,678.19Supplier Number: 301971
- TetraCon 325-15Graphite15.3 mm (5/8")15 m (50 ft.)Epoxy0.475 cm⁻¹Discontinued-Supplier Number: 301973
- TetraCon® VEpoxy15.3 mm (5/8")1.5 m (5 ft.)Epoxy0.475 cm⁻¹Discontinued-Supplier Number: -
- TetraCon 325Graphite15.3 mm (5/8")1.5 m (5 ft.)Epoxy0.475 cm⁻¹Log in for availabilityEach$2,526.96Supplier Number: 301960