Spectra/Por® 6 Dialysis Membranes, MWCO 1000 to 50000, Spectrum® Laboratories
Available in wet form in 0.05% sodium azide as a preservative only.
- Ready to use
- Should be refrigerated
- Transparent and flexible
Chemically altered to offer more MWCO. Membranes contain nominal quantities of contaminants.
: Roll length: 10 m (32.8'). Trial kit roll length: 1 m (3.28").
Frequently Bought Together