Hazardous Material Shippers, HDPE, UN Marked, Qorpak®
Supplier: Berlin Packaging
Certificates
About this item
Four-pack multiple convenience packs suitable for shipping hazardous materials.
- Narrow mouth bottles for liquids
- Wide mouth bottles for solid and viscous materials
Certified packaging is supplied complete with high density polyethylene container, polypropylene closure, corrugated outer packaging, cushioning material, absorbent, and tape.
Meet UN 4G requirements.
Warning: Shipper must not make any substitutions. Failure to heed this warning would invalidate package compliance.
Frequently Bought Together