MOPS Minimal Medium Kit, Teknova
MOPS Minimal Medium Kit, Teknova
MOPS Minimal Medium Kit
Catalog # CA100218-936
Supplier:   Teknova
Supplier #:  M2106
MOPS Minimal Medium Kit, Teknova
Catalog # CA100218-936
Supplier:  Teknova
CAS Number:  
MOPS Minimal Medium Kit, Teknova
MOPS Minimal Medium Kit
CAS Number:  
Price
$512.49
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Designed for the growth of E
MOPS mixture should be stored frozen. The potassium phosphate dibasic solution and 20% glucose solution should be stored at room temperature.
coli and other enterobacteria. Kit consists of 100mL 10X MOPS mixture (200059-678), 10mL 0.132M potassium phosphate diabase solution (200059-652), and 100mL 20% glucose solution (100216-728), and makes 5 liters.
Specifications
Specifications
  • Désignation:
    MOPS Minimal Medium Kit