VWR® Contour™ Top-Mounted Apron Legs
Fournisseur: VWR International
Certificats
À propos de cet article
Each 5.1 cm (2") square leg is welded to 10.2 cm (4") square steel plate.
- Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
- No ragged edges
- Light neutral color
The complete apron is used to create a knee space for a desk area or to create a free-standing table. Apron components are sold separately.
Frequently Bought Together