SP Bel-Art Poxygrid® Petri Dish and Contact Plate Racks, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Ideal for organizing laboratory environments, these welded steel wire racks are available in multiple sizes and configurations.
- Facilitates transport and dish incubation
- Epoxy-coated steel wire construction
- Suitable for 60 mm contact plates
- Durable
- Autoclavable
Designed for convenient organization, racks are useful for transport, incubation, and storage of petri dishes or contact plates. Racks are steam autoclavable at 121°C (250°F).
Frequently Bought Together