polyproLABS® Sink Base Cabinets, Air Control
Corrosion-resistant casework cabinets are constructed of stress-relieved, acid-resistant, 1.3 cm (½") thick polypropylene
- No metals are used in cabinets
Joints are thermally welded by a special process that ensures leak-tight, flush unions. All hardware, including door hinges, screws, latches, and handles, is constructed of polypropylene. . Toe space measures 10.2H×7.6D cm (4×3"). All cabinets have an open back for plumbing access and two doors made from solid 1cm (⅜") thick polypropylene with beveled edges.
Frequently Bought Together