ContactUsLinkComponent
SP Bel-Art Contact Plate Rack, 72 mm, Bel-Art Products, a part of SP
Catalog # 12000-068
undefined
SP Bel-Art Contact Plate Rack, 72 mm, Bel-Art Products, a part of SP
Catalog # 12000-068
Supplier Number:  F18990-0000
Specifications
  • Capacity:
    60 Plates/Dishes
  • Description:
    Contact Plate Rack
  • Dimensions:
    26.7L×16.5W×17.8H cm (101/2 x 61/2 x 7")
  • Cat. no.:
    12000-068
  • Supplier No.:
    189900000
Specifications
About this item
Safely transport, incubate, and store up to 60 contact plates with lids up to 72 mm in diameter (RODAC®, Becton Dickinson) and streamline benchtop sample processing.
  • Polypropylene construction
  • Autoclavable
  • Quick and easy to assemble
  • Built-in handle
  • Ample labeling space
Rack is configured in 6 columns with bottom access slots for easy removal of plates from the rack.
Center divider has a built-in handle and ample space for labeling. Racks interlock for safe stacking up to four high. Made from polypropylene, rack is autoclavable at 121 °C (250 °F). Quick assembly.