SP Bel-Art Petri Dish Rack, 100 mm, Clear TPX® Body, Bel-Art Products, a part of SP
Catalogue # 47751-754
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Capacity:54 Dishes/Plates
- Dimensions:34L x 20W x 23H cm (131/2 x 8 x 91/4")
- Cat. no.:47751-754
- Supplier No.:189850100
Spécifications
À propos de cet article
Clear, TPX® rack with polypropylene posts securely holds up to fifty-four 100 mm Petri dishes.
- Clear body
- TPX® construction with polypropylene posts
- Shipped flat and easy to assemble
Easily viewed through the clear base and divider, dishes can be removed with one hand.
Built-in handle provides easy transport, and rubber feet help prevent rack from sliding on the benchtop. Racks are shipped flat and can be assembled in minutes.