SP Bel-Art Poxygrid® Petri Dish Dispensing Racks, 100 mm , Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Petri dishes are easily dispensed one at a time from the bottom of the stack.
- Tilts toward the rear for convenient presentation of dishes
- Not suitable for glass petri dishes
Each column holds twenty 10 x 2 cm (nominal size) plastic petri dishes per stack, which are loaded at the top.
Frequently Bought Together