VWR® Inoculating Turntables | Avantor
VWR® Inoculating Turntables
VWR® Inoculating Turntables
Supplier:  VWR International
Certificates
VWR® Inoculating Turntables
VWR® Inoculating Turntables
2 Options Available Below
Product Details & Documents
Hand-operated inoculating turntables produce almost concentric circles of bacterial colonies evenly distributed across Petri dishes.
  • Cast iron construction
  • Gray baked acrylic enamel
  • Now with Best-in-Class, Exclusive 5-Year Warranty
The small inoculating turntable accommodates 100 mm Petri dishes. This 76.2 mm (3") high turntable has a tripod base to bring work to approximately eye level. The center disk is covered with nonskid rubber.
The large inoculating turntable accommodates 100 mm or 150 mm Petri dishes. This 32 mm (11/4") high turntable is ideal for steadying forearm on the work surface while plating. It contains two circular sections: a top section that has a raised gripper lip, and a recessed center disk that is covered with nonskid rubber.
Cast iron turntables are coated with gray baked acrylic enamel.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Small turntable (with pedestal)
    114.3Ø×76.2 mm H (4½×3")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $547.99
    $547.99
    Prix promotionnel pour les nouveaux clients
    Each
       $547.99
  • Large turntable (without pedestal)
    150Ø×32 mm H (529/32×1¼")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $683.74
    $683.74
    Prix promotionnel pour les nouveaux clients
    Each
       $683.74