Case systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
- The key component of case systems casework is the plastic laminate
- Plastic laminate provides superior resistance to chemicals, scratching, and abrasion
- The unique hardware system provides rigid support for vertical integration without limiting adjustability
A perfect choice for laboratory environments. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.
The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards. Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware. The result cabinets that are as attractive as they are durable.
Case cabinetry gives you the adaptability necessary in today’s laboratories. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves.
Cabinets have eight drawers. Sitting height base cabinets are common for traditional laboratory and technical environments. They are usually installed for permanent or semi-permanent layouts. Color: Beige.