Mobile Wire Security Units, Metro™
Supplier:   Metropolitan Wire
Mobile Wire Security Units, Metro™
Supplier:  Metropolitan Wire
Certificates
Mobile Wire Security Units, Metro™
Mobile Wire Security Units, Metro™
35 Options Available Below
Product Details & Documents
Security units are designed to protect materials and sensitive items from loss or pilferage
Note: Given height is for unit without a Metro dolly or casters.
Units move easily to allow thorough cleaning or inspection, and they provide the advantage of quick, protective transportation. Heavy-gauge, open wire construction allows contents to be visible at all times, making inspection and retrieval quick and simple. An ergonomic, 1/4-turn door handle simplifies opening and closing. Each unit is equipped with a set of double doors that open 270° and can be secured along the sides of the unit. Super Erecta* and qwikSLOT* shelf designs are easily adjustable and allow for flexibility to meet changing needs. Super Erecta shelves attach to posts via tapered sleeves; qwikSLOT shelves rest securely on support clips that can be quickly and easily repostitioned. Shelves can be positioned in 2.5cm (1") increments along the entire height of the posts. Security units are available in chrome, stainless steel, or Metroseal 3, which provides Microban* antimicrobial product protection. All units are shipped knocked-down and can be quickly and easily assembled right from packaging.
 : Chrome and Metroseal 3 units are supplied with 12.7cm (5") standard-duty polyurethane-tread stem casters. Stainless steel units do not include casters; see 19766-236 series to purchase them. Contact your Avantor representative for the proper caster for your application. For all units, interior shelves must be purchased separately. For Super Erecta shelves, see 22233-716 series; for qwikSLOT shelves, contact your Avantor representative. For stationary wire security units, see 82023-452 series. Heavy-duty stainless steel models include staked posts for use with standard Metro dollies. Dolly bases and plate casters must be ordered separately; contact your Avantor representative for more information.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Largeur
    Hauteur
    Finish
    Casters
    Shipping Weight
    Availability
    Price
  • Super Erecta Security Unit
    40¾"
    27¼"
    62"
    Stainless steel
    137 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11,551.39
    $11,551.39
    Each
       $11,551.39
    Supplier Number: SEC53S-SD
  • Super Erecta Security Unit
    52¾"
    27¼"
    62"
    Stainless steel
    162 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14,079.41
    $14,079.41
    Each
       $14,079.41
    Supplier Number: SEC55S-SD
  • Super Erecta Security Unit
    65"
    27¼"
    62"
    Stainless steel
    178 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14,571.07
    $14,571.07
    Each
       $14,571.07
    Supplier Number: SEC56S-SD
  • Super Erecta Security Unit
    40¾"
    27¼"
    68½"
    Chrome
    2 swivel, 2 brake
    162 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,094.78
    $5,094.78
    Each
       $5,094.78
    Supplier Number: SEC53EC
  • Super Erecta Security Unit
    52¾"
    27¼"
    68½"
    Chrome
    2 swivel, 2 brake
    182 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,165.71
    $6,165.71
    Each
       $6,165.71
    Supplier Number: SEC55EC
  • Super Erecta Security Unit
    65"
    27¼"
    68½"
    Chrome
    2 swivel, 2 brake
    203 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,582.73
    $6,582.73
    Each
       $6,582.73
    Supplier Number: SEC56EC
  • Super Erecta Security Unit
    40¾"
    27¼"
    68½"
    Metroseal 3
    4 swivel
    162 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,968.75
    $5,968.75
    Each
       $5,968.75
    Supplier Number: SEC53EK3
  • Super Erecta Security Unit
    52¾"
    27¼"
    68½"
    Metroseal 3
    4 swivel
    182 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,227.72
    $7,227.72
    Each
       $7,227.72
    Supplier Number: SEC55EK3
  • Super Erecta Security Unit
    65"
    27¼"
    68½"
    Metroseal 3
    4 swivel
    203 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,777.55
    $7,777.55
    Each
       $7,777.55
    Supplier Number: SEC56EK3
  • qwikSLOT Security Unit
    40¾"
    27¼"
    68½"
    Chrome
    2 swivel, 2 brake
    162 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,308.39
    $5,308.39
    Each
       $5,308.39
    Supplier Number: SEC53ECQ
  • qwikSLOT Security Unit
    52¾"
    27¼"
    68½"
    Chrome
    2 swivel, 2 brake
    182 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,402.94
    $6,402.94
    Each
       $6,402.94
    Supplier Number: SEC55ECQ
  • qwikSLOT Security Unit
    65"
    27¼"
    68½"
    Chrome
    2 swivel, 2 brake
    203 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,839.74
    $6,839.74
    Each
       $6,839.74
    Supplier Number: SEC56ECQ
  • Super Erecta Security Unit
    40¾"
    27¼"
    68½"
    Metroseal 3
    2 swivel, 2 brake
    162 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,919.96
    $5,919.96
    Each
       $5,919.96
    Supplier Number: SEC53DK3
  • Super Erecta Security Unit
    52¾"
    27¼"
    68½"
    Metroseal 3
    2 swivel, 2 brake
    182 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,178.86
    $7,178.86
    Each
       $7,178.86
    Supplier Number: SEC55DK3
  • Super Erecta Security Unit
    65"
    27¼"
    68½"
    Metroseal 3
    2 swivel, 2 brake
    203 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,721.02
    $7,721.02
    Each
       $7,721.02
    Supplier Number: SEC56DK3
  • qwikSLOT Security Unit
    40¾"
    27¼"
    68½"
    Chrome
    4 swivel
    162 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,267.21
    $5,267.21
    Each
       $5,267.21
    Supplier Number: SEC53DCQ
  • qwikSLOT Security Unit
    52¾"
    27¼"
    68½"
    Chrome
    4 swivel
    182 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,363.96
    $6,363.96
    Each
       $6,363.96
    Supplier Number: SEC55DCQ
  • qwikSLOT Security Unit
    65"
    27¼"
    68½"
    Chrome
    4 swivel
    203 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,811.83
    $6,811.83
    Each
       $6,811.83
    Supplier Number: SEC56DCQ
  • Super Erecta Security Unit
    40¾"
    21½"
    68½"
    Chrome
    2 swivel, 2 brake
    146 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,984.59
    $4,984.59
    Each
       $4,984.59
    Supplier Number: SEC33EC
  • qwikSLOT Security Unit
    40¾"
    21½"
    68½"
    Chrome
    2 swivel, 2 brake
    146 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,208.03
    $5,208.03
    Each
       $5,208.03
    Supplier Number: SEC33ECQ
  • Super Erecta Security Unit
    40¾"
    21½"
    68½"
    Metroseal 3
    2 swivel, 2 brake
    146 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,849.08
    $5,849.08
    Each
       $5,849.08
    Supplier Number: SEC33EK3
  • Super Erecta Security Unit
    40¾"
    21½"
    62"
    Stainless steel
    138 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11,142.09
    $11,142.09
    Each
       $11,142.09
    Supplier Number: SEC33S-SD
  • Super Erecta Security Unit
    52¾"
    21½"
    68½"
    Chrome
    2 swivel, 2 brake
    165 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,003.54
    $6,003.54
    Each
       $6,003.54
    Supplier Number: SEC35EC
  • qwikSLOT Security Unit
    52¾"
    21½"
    68½"
    Chrome
    2 swivel, 2 brake
    165 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,268.67
    $6,268.67
    Each
       $6,268.67
    Supplier Number: SEC35ECQ
  • Super Erecta Security Unit
    52¾"
    21½"
    68½"
    Metroseal 3
    2 swivel, 2 brake
    165 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,074.45
    $7,074.45
    Each
       $7,074.45
    Supplier Number: SEC35EK3
  • Super Erecta Security Unit
    52¾"
    21½"
    62"
    Stainless steel
    157 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13,543.99
    $13,543.99
    Each
       $13,543.99
    Supplier Number: SEC35S-SD
  • Super Erecta Security Unit
    40¾"
    33½"
    68½"
    Chrome
    2 swivel, 2 brake
    175 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,376.80
    $6,376.80
    Each
       $6,376.80
    Supplier Number: SEC63EC
  • Super Erecta Security Unit
    40¾"
    33½"
    62"
    Stainless steel
    167 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14,442.43
    $14,442.43
    Each
       $14,442.43
    Supplier Number: SEC63S-SD
  • Super Erecta Security Unit
    52¾"
    33½"
    68½"
    Chrome
    2 swivel, 2 brake
    202 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,674.36
    $7,674.36
    Each
       $7,674.36
    Supplier Number: SEC65EC
  • Super Erecta Security Unit
    52¾"
    33½"
    62"
    Stainless steel
    193 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17,580.74
    $17,580.74
    Each
       $17,580.74
    Supplier Number: SEC65S-SD
  • Super Erecta Security Unit
    65"
    33½"
    68½"
    Chrome
    2 swivel, 2 brake
    223 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,245.75
    $8,245.75
    Each
       $8,245.75
    Supplier Number: SEC66EC
  • Super Erecta Security Unit
    65"
    33½"
    62"
    Stainless steel
    215 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18,201.07
    $18,201.07
    Each
       $18,201.07
    Supplier Number: SEC66S-SD
  • Super Erecta Security Unit
    38½"
    28¹⁄₁₆"
    62"
    Stainless steel, heavy-duty
    154 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11,551.39
    $11,551.39
    Each
       $11,551.39
    Supplier Number: SEC53S-HD
  • Super Erecta Security Unit
    50½"
    28¹⁄₁₆"
    62"
    Stainless steel, heavy-duty
    174 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14,079.41
    $14,079.41
    Each
       $14,079.41
    Supplier Number: SEC55S-HD
  • Super Erecta Security Unit
    63"
    28¹⁄₁₆"
    62"
    Stainless steel, heavy-duty
    195 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14,571.07
    $14,571.07
    Each
       $14,571.07
    Supplier Number: SEC56S-HD