Nunc® UpCell® MultiDishes, Thermo Scientific
Single-use vented multidishes feature a temperature-responsive cell culture surface that is ideal for culture passaging, single-cell analyses, and cell transplantation research
The surface allows harvesting of cells with intact surface proteins. Adherent cells are released by reducing the temperature of the cell culture. High cell viability is achieved with minimal hands-on time and without trypsinization or physical force. Ideal for harvesting of cell sheets and creations of 3-D tissue models held together by normal cell junctions and the extracellular matrix. Sterile.
Frequently Bought Together