ContactUsLinkComponent VWR® Racks and Replacement Parts for VWR Signature™ Ultra-Low Temperature and VWR® Low-Temperature and Ultra-Low Temperature Freezers
VWR® Racks and Replacement Parts for VWR Signature™ Ultra-Low Temperature and VWR® Low-Temperature and Ultra-Low Temperature Freezers
VWR® offers a complete line of accessories for Low-Temperature and Ultra-Low Temperature freezers. Choose the accessories that will meet your laboratory’s needs and safely store your frozen samples.
Sensaphone telephone dialing systems connect to remote alarm contacts on VWR® Signature and VWR® freezers
Chart recorders are 15.2cm (6") circular models with 7-day temperature recording capability. They are for use with all 120V and 230V freezers. Available data loggers serve as a paperless version of a chart recorder.
The 4-channel system monitors up to four inputs and dials up to four phone numbers until the warning message is received and acknowledged. The 8-channel system monitors up to eight inputs and dials up to eight phone numbers until the warning message is received and acknowledged. Battery back-up is standard. Six D-size alkaline batteries are required for backup.
Ordering information: Some accessories are factory-installed and must be specified when ordering. For details on ordering these products, contact your Avantor sales representative. Deluxe CO2 and LN2 backup systems include hose and necessary fittings. Sample probe kits include probe clip and probe. For VWR® Signature ULT Freezers, see series 14230-110; for VWR® Low-Temperature Upright and ULT Upright and Chest Freezers, see 14230-144 series. IQ/OQ Validation services are available, contact your Avantor sales representative for further information.