Nunclon™ Delta MultiDish Inserts, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes
Use for transport studies, toxicity tests, chemotaxis studies, and electron microscopy. They are also suitable for phase contrast and Nomarski optic systems. Inserts have been treated and tested for cell attachment, providing easy cultivation of most cell types without the need for matrix coating. They feature tabs for better handling. Nontoxic. Nonpyrogenic.
Ordering information: Packed in resealable bags.
Frequently Bought Together