Synthware Sure-Grip Separatory Funnels
À propos de cet article
Globe-shaped separatory funnel features a long stopper neck and a straight section between the funnel body and stopcock that provides a secure grip while shaking
- Sure-grip design
- Heavy wall borosilicate construction
- Ground glass joint
The straight section also aids in clamping. Available with PTFE stopcocks. Funnels are supplied complete with one PTFE or hollow glass stopper.
5 Options Available Below
Product Details & Documents
Globe-shaped separatory funnel features a long stopper neck and a straight section between the funnel body and stopcock that provides a secure grip while shaking
- Sure-grip design
- Heavy wall borosilicate construction
- Ground glass joint
The straight section also aids in clamping. Available with PTFE stopcocks. Funnels are supplied complete with one PTFE or hollow glass stopper.
Product Family Options
Informations sur le produit
- DésignationCapacitéStopcock SizeStopper No.AvailabilityPrice
- Supplier Number: F687500A
- Supplier Number: F687000A
- Supplier Number: F687000
- Supplier Number: F68272LA
- Supplier Number: F68272L