Falcon® Organ Culture Dish, Sterile, Corning
CA25381-141
: Corning
- Ø:60 mm
- Description:Organ Culture Dish, Center Well Area 2.89 cm2
- Dimensions:54.84 O.D. x 13.56H mm Actual
- Height:15 mm
- Cat. no.:CA25381-141
- Supplier No.:353037
Designed for tissue and organ culture techniques
Polystyrene dish is tissue culture treated by vacuum-gas plasma. Sterilized by gamma irradiation. Nonpyrogenic.
: Packaged 20 per sleeve.