ContactUsLinkComponent
S-Klenz® Scale Remover
Catalog # 77531-584
Supplier:  Steris
New Product
S-Klenz® Scale Remover
Catalog # 77531-584
Supplier:  Steris
Supplier Number:  1J8108
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Scale remover
  • Size:
    4×1 gal.
  • Cat. no.:
    77531-584
  • Packaging:
    Plastic bottle
  • Form:
    Liquid
Specifications
About this item
S-Klenz Scale Remover is a phosphoric acid-based film and scale remover. This instrument rust inhibitor is also used for de-scaling equipment.
  • Phosphoric acid-based de-scaler effectively removes scale deposits
  • Free-rinsing surgical instrument scale remover leaves no residue behind in automated washer/disinfectors
  • Easily measured and dispensed liquid formula
Effective at removing lime scale, urine scale, and corrosion from stainless steel, glass, and all plastics. The wetting and penetrating agents of the formula remove film and scale, but is not recommended for use on aluminum surfaces, galvanized metals, and mild steel.