ContactUsLinkComponent
Synthware Tube, Drying, 90°, Microscale, Kemtech America
Catalog # 60004-132
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Tube, Drying, 90°, Microscale, Kemtech America
Catalog # 60004-132
Supplier:  Kemtech America
Supplier Number:  A851410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • ST Joint No.:
    14/10
  • Cat. no.:
    60004-132
  • Supplier No.:
    A851410
Specifications
About this item
Drying tube with a 90° angle.
  • Heavy wall borosilicate construction
  • Ground glass joint