Synthware Tubes, Craig Recrystallization, Microscale, Kemtech America
Catalogue # 60004-148
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:2 mL, Complete
- Cat. no.:60004-148
- Supplier No.:T140002
Spécifications
À propos de cet article
Craig recystalllization tubes for microscale applications.
- 3.3 Expansion borosilicate glass
- Complete set
Complete tube includes a glass outer tube and an inner plug.