InLab® Versatile Pro 3-in-1 pH Electrode, Replaceable Junction, METTLER TOLEDO®
Catalog # CA97001-656
Supplier: Mettler Toledo
Specifications
- Temperature Range:0...100 °C
- Length:120 mm
- Cat. no.:CA97001-656
- pH Range:0 - 14
- Body Material:Polysulfone
- Internal Reference:ARGENTHAL
- Junction:Ceramic
- Fill Solution:3M KCl
- Supplier No.:51343031
Specifications
About this item
Electrode features a replaceable ceramic junction, which offers a solution to contaminated junctions without the higher cost or cleaning efforts required with open or sleeve junctions
- Replaceable ceramic junction
- Integrated temperature probe
- Polysulfone shaft, universal glass membrane
- ARGENTHAL™ lead-off and silver ion trap
- BNC/RCA (Cinch) connector
Electrode also features integrated NTC 30 k[OMEGA] temperature probe, robust polysulfone shaft, universal glass membrane with low resistance, ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures, and BNC/RCA (Cinch) connector.