SICURIX® Ribbon Cartridge
Catalogue # 77531-646
Fournisseur: Janitorial Supplies
New Product
Spécifications
- For Use With:Fargo® DTC1000, DTC1250e
- Color:YMCKO
- Description:45000 printer ribbon cartridge, 8.13×10.40×6.02" (W×H×D), 2.875 lbs.
- Cat. no.:77531-646
Spécifications
À propos de cet article
The SICURIX® ribbon cartridge is an alternative for Fargo (45000). Full-color ribbon cartridge is especially designed for printing a combination of full-color images and one-color text or bar codes in the Fargo DTC1000 Card Printer/Encoder. The Fargo 45000 color ribbon features a resin black and clear overlay panel and includes a built-in cleaning roller. YMCKO ribbon yields 250 images.
- Designed for use with the Fargo DTC1000 Card Printer/Encoder
- Features a resin black and clear overlay panel
- Includes a built-in cleaning roller
Print technology: Dye sublimation and thermal transfer