Microcentrifuge Tubes with Screw Cap, Molecular BioProducts
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
Manufactured from high quality polypropylene, these clear microcentrifuge tubes are designed for vapor phase liquid nitrogen storage, boiling, and autoclaving procedures.
- RNase-, DNase-, and endotoxin-free
- Nonsterile
Tubes are sturdy enough to withstand centrifugation at 20000×g. All tubes are supplied with unattached screw cap and O-ring. Conical bottom tubes allow for more complete sample recovery. Freestanding tubes feature external ribs that assist in positioning and locking tubes into racks, allowing for convenient one-handed opening and closing.
Spécifications
- Catalog No:
- 89233-704
- 89047-094
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)