Microcentrifuge Tubes, Polypropylene, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Microcentrifuge tubes are manufactured from highly transparent, inert polypropylene resin that offers superior chemical resistance and thermal stability.
- Polypropylene construction
- Highly transparent and inert
- Superior chemical and temperature resistance
- Autoclavable
- Available nonsterile or sterile
Color-coded tubes are available for easy and quick identification.
Tubes are designed to suit most analyzer systems, including Eppendorf®, Viatron, Roche®, and Beckman® systems.
All 1.5 ml tubes feature molded graduations. Ideal solution for secure, micro-volume liquid storage. Options for conical, skirted bottom available. Tubes are nontoxic and nonpyrogenic.
Frequently Bought Together