Costar® Low Binding Microcentrifuge Tubes, Polypropylene, Corning
Catalog # 29442-578
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:0.65 ml
- Cat. no.:29442-578
- Maximum RCF:17000×g
- Supplier No.:3206
Specifications
About this item
Low binding microcentrifuge tubes feature a bonded polymer technology that reduces protein and nucleic acid binding, resulting in better sample recovery.
With external graduations and frosted writing spot for easy sample identification. Tubes are disposable and autoclavable. Color: natural.
Certifications: Certified RNase- and DNase-free.