Pierce™ Slide-A-Lyzer® MINI Dialysis Units, Thermo Scientific
Versatile, disposable units are ideal for sample preparation to dialyze proteins or nucleic acids.
- Quick and convenient
- Durable, with disposable PP caps
- Plastic construction
Device minimises sample loss compared to filtration and resin systems. Performs desalting of nucleic acids and proteins in less than 15 minutes, which improves sample resolution, quantification and conjugation.
Frequently Bought Together