Test Tubes, Greiner Bio-One
Test Tubes, Greiner Bio-One
Test Tubes, Greiner Bio-One
Test Tubes, Greiner Bio-One
Test Tubes, Greiner Bio-One
These high-quality test tubes are available in a wide variety of materials, sizes, and volume ranges.
  • Excellent optical clarity
  • Round, flat, or conical bottom designs available
  • Polycarbonate, polypropylene, or polystyrene construction available
  • Exceptional thermal and chemical stability
Test tubes provide excellent optical clarity, and are available in round or conical bottom designs. Unless otherwise noted, tube caps are not included.
Ordering information: Test tube 82050-248 is supplied sterile.
Documents
Product Family Options
Polypropylene Tubes with Round Bottoms
  • Capacité
    Désignation
    Hauteur
    Ø
  • 4 ml
    Without Cap
    55 mm
    12 mm
    Case of 3,600
    Case of 3,600 - $362.00
    $362.00
    Case of 3,600
       $362.00
     : 112201
  • 12 ml
    Without Cap
    100 mm
    16 mm
    Case of 1,600
    Case of 1,600 - $388.00
    $388.00
    Case of 1,600
       $388.00
     : 160201
  • 12 ml
    Without Cap
    100 mm
    16.8 mm
     : 163201
  • 14 ml
    Without Cap
    100 mm
    16 mm
     : -
  • 14 ml
    Without Cap
    100 mm
    17 mm
    Case of 1,500
    Case of 1,500 - $368.00
    $368.00
    Case of 1,500
       $368.00
     : 187201
Polystyrene Tubes with Round Bottoms
  • Capacité
    Désignation
    Hauteur
    Ø
  • 2 ml
    Without Cap
    40 mm
    10.5 mm
     : 103101
  • 5 ml
    Without Cap
    75 mm
    12 mm
    Case of 2,000
    Case of 2,000 - $198.09
    $198.09
    Case of 2,000
       $198.09
     : 115101
  • 6.5 ml
    Without Cap
    65 mm
    16 mm
     : -
  • 10 ml
    Without Cap
    100 mm
    14 mm
    Case of 1,400
    Case of 1,400 - $236.00
    $236.00
    Case of 1,400
       $236.00
     : 136101
  • 12 ml
    Without Cap
    100 mm
    16 mm
    Case of 1,600
    Case of 1,600 - $209.00
    $209.00
    Case of 1,600
       $209.00
     : 160101
  • 14 ml
    With Black Screw Cap
    100 mm
    16 mm
     : -
  • 12 ml
    With Cap
    100 mm
    16 mm
     : 163177
  • 14 ml
    Without Cap
    100 mm
    17 mm
    Case of 1,500
    Case of 1,500 - $248.20
    $248.20
    Case of 1,500
       $248.20
     : 187101
  • 15 ml
    With Blue Screw Cap
    120 mm
    17 mm
     : 186171
  • 20 ml
    Without Cap
    152 mm
    16 mm
    Case of 1,500
    Case of 1,500 - $662.00
    $662.00
    Case of 1,500
       $662.00
     : 169101
Polystyrene Tubes with Conical Bottoms
  • Capacité
    Désignation
    Hauteur
    Ø
  • 12 ml
    With Blue Bayonet Cap
    100 mm
    16.8 mm
     : 164161
  • 15 ml
    With Blue Screw Cap
    120 mm
    17 mm
    Case of 500
    Case of 500 - $599.70
    $599.70
    Case of 500
       $599.70
     : 188161
  • 15 ml
    With Blue Screw Cap
    120 mm
    17 mm
    Case of 1,000
    Case of 1,000 - $1,202.09
    $1,202.09
    Case of 1,000
       $1,202.09
     : 188171
Polypropylene Tubes with Two Position Vent Stopper
  • Capacité
    Désignation
    Hauteur
    Ø
  • 5 ml
    Graduated Tube with Snap Cap
    75 mm
    12 mm
     : -
  • 5 ml
    Graduated Tube with Snap Cap
    75 mm
    12 mm
    Case of 2,000
    Case of 2,000 - $1,030.00
    $1,030.00
    Case of 2,000
       $1,030.00
     : 115262
  • 12 ml
    With Snap Cap
    77 mm
    17 mm
     : -
  • 14 ml
    With Snap Cap
    95 mm
    18 mm
     : -
  • 14 ml
    With Snap Cap
    95 mm
    18 mm
    Case of 1,000
    Case of 1,000 - $535.00
    $535.00
    Case of 1,000
       $535.00
     : 187262
Polystyrene Tubes with Two Position Vent Stopper
  • Capacité
    Désignation
    Hauteur
    Ø
  • 4.5 ml
    With Snap Cap
    75 mm
    12.4 mm
     : -
  • 4.5 ml
    With Snap Cap
    75 mm
    12.4 mm
    Case of 2,000
    Case of 2,000 - $931.96
    $931.96
    Case of 2,000
       $931.96
     : 120180
  • 14 ml
    With Snap Cap
    95 mm
    18 mm
     : -
  • 14 ml
    With Snap Cap
    95 mm
    18 mm
    Case of 1,000
    Case of 1,000 - $610.00
    $610.00
    Case of 1,000
       $610.00
     : 191180
Accessories
  •  
    Description
  • 10066-772
    Screw Cap for Polypropylene Tube, White, For Model 163201
    Case of 9,000 - $935.15