Corning® Culture Tube, Disposable, Polystyrene, Sterile | Avantor
Corning® Culture Tube, Disposable, Polystyrene, Sterile
   Options durables disponibles
Supplier:   Corning
Corning® Culture Tube, Disposable, Polystyrene, Sterile
Supplier:  Corning
   Options durables disponibles
Certificates
Corning® Culture Tube, Disposable, Polystyrene, Sterile
Corning® Culture Tube, Disposable, Polystyrene, Sterile
2 Options Available Below
Product Details & Documents
Optically clear PS.
  • TC treated tubes supplied racked
  • Untreated tubes provided bulk packed
  • Sterilized by gamma radiation
  • Certified non-pyrogenic
Culture tubes with threaded plug seal caps.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 60818-601
  • 62404-982
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Hauteur
    Ø
    Availability
    Price
  • 10 ml
    Disposable sterile polystyrene culture tube
    125 mm
    16 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $721.65
    $721.65
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $721.65
    Supplier Number: 430157
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    60818-601
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 10 ml
    Disposable sterile polystyrene culture tube
    125 mm
    16 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $742.90
    $742.90
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $742.90
    Supplier Number: 430172
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    62404-982
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact