Drop In Sink, Self Rim Design with No Deck, One Bowl
Fournisseur: Eagle Group
New Product
Certificats
À propos de cet article
Sinks can be mounted onto top or bottom of countertop. Heavy gauge type 300 series stain less steel coved bowls with large radius.
- All sinks feature 35 mm (¹³/₈") trim on all four sides of sink
- 121 mm (⁴³/₄") sump in sink bowl
- 89 mm (³¹/₂") basket drain with 38 mm (¹¹/₂") outlet
- Self rimming
16, 18 or 20 gauge industrial grade construction and quality. 16 gauge bowls are fabricated, 18 and 20 gauge bowls are deep-drawn.
Frequently Bought Together