Nunc® Disposable Tubes, Round-Bottom, Polystyrene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These transparent, polystyrene tubes offer a high degree of thermal and chemical stability.
Tubes withstand centrifuged forces up to 3000 ×g and are suitable for sample transport or the culture of microorganisms.
Ordering information: Additional polyethylene stoppers are sold separately.
Delivery: Tube 62409-465 is sterilized and supplied with a mounted stopper.
Frequently Bought Together