InLab® 605 Dissolved Oxygen Sensors, METTLER TOLEDO®
Sensors ensure a high degree of accuracy and reproducibility for polarographic dissolved oxygen determination.
- High degree of accuracy
- For polargraphic DO determination
- Withstands harsh conditions
- Steel mesh protects membrane
- Reinforced shaft
Designed to withstand the harshest of conditions, sensors feature a membrane protected by steel mesh, a shaft constructed from glass fiber reinforced polyphenylene sulfide (PPS), and BNC/Cinch connectors. For use with SevenGo® portable dissolved oxygen meters. Sensors meet full IP67 protection standards.
: Sensors with ISM® technology also available. Accessories sold separately.
Frequently Bought Together