PYREXPLUS® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopper, Corning
 :   Corning
PYREXPLUS® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopper, Corning
 :  Corning
PYREXPLUS® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopper, Corning
PYREXPLUS® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopper, Corning
PYREXPLUS®, borosilicate glass.
  • Fluoropolymer resin stopcock reduces freezing problems and grease contamination
  • Coating is autoclavable to 121 °C (250 °F) moist heat or 110 °C (230 °F) dry heat
  • Resistant to thermal shock
PYREXPLUS® labware allows for a better hand grip and clamp hold. PVC coating makes cleanup faster and easier by reducing spill volume if breakage occurs. Can be written on with any standard lab marker, permanent or water-soluble. Do not use plastic-coated ware over direct or open flame.
  • Catalog No:
  • 89091-670
  • 89091-696
  • 30356-249
  • 30356-252
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Product Family Options
  • Capacité
    Stopcock Size
    Stopper No.
  • 125 ml
    2
    22
    Case of 4
    Case of 4 - $1,800.00
    $1,800.00
    Case of 4
       $1,800.00
     : 66402-125
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89091-670
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 ml
    4
    27
    Case of 4
    Case of 4 - $2,420.00
    $2,420.00
    Case of 4
       $2,420.00
     : 66402-500
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89091-696
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1000 ml
    4
    27
    Case of 1
    Case of 1 - $862.00
    $862.00
    Case of 1
       $862.00
     : 66402-1L
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30356-249
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2000 ml
    6
    38
    Case of 1
    Case of 1 - $1,120.00
    $1,120.00
    Case of 1
       $1,120.00
     : 66402-2L
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    30356-252
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact