PYREX® Separatory Funnels, Squibb, Pear-Shape, Fluoropolymer Resin Stopcock, Corning
Fournisseur: Corning
Certificats
À propos de cet article
PYREX® Borosilicate glass.
With PTFE stopcock and linear, high-density, closed-bottom polyethylene stoppers which conform to [ST] stopper dimensions.
Spécifications
- Catalog No:
- 89090-654
- 89090-792
- 89091-508
- 89090-880
- 30356-700
- 30356-722
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey