Ward's® NYS Investigations Pressure Block Set
470356-614
New Product
- Description:Pressure block set
- Cat. no.:470356-614
Corresponds to the NYS earth and pace science investigation: The sky is the limit.
- Ready to go - no assembly required
- Reusable
- Stackable
- Block sets are approximately equal masses
This set consists of 75 red blocks and 75 blue blocks. The red blocks are cube with sides approximately 1³/₈". The blue blocks have the same dimensions except are half the height.