ASTM Class 7 Hooked Weights
Supplier: Troemner
Jump to:
À propos de cet article
ASTM hooked weights for weighing operations in physical and chemical classrooms, such as force measuring.
- 303 stainless steel
- ASTM Class 7
All surfaces are satin finish and meet or exceed ANSI/ASTM E617 tolerance and design restrictions.
7 Options Available Below
Product Details & Documents
ASTM hooked weights for weighing operations in physical and chemical classrooms, such as force measuring.
- 303 stainless steel
- ASTM Class 7
All surfaces are satin finish and meet or exceed ANSI/ASTM E617 tolerance and design restrictions.
Product Family Options
Informations sur le produit
- WeightAvailabilityPrice
- Supplier Number: 30390784
- Supplier Number: 30390783
- Supplier Number: 30390782
- Supplier Number: 30390781
- Supplier Number: 30390780
- Supplier Number: 30390779
- Supplier Number: 30390778