ASTM Class 4 Precision Weight Sets
Supplier: Mettler Toledo
Jump to:
About this item
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
- 316 stainless steel
- Low magnetic permeability
- High corrosion resistance
- High gloss polish
- ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
14 Options Available Below
Product Details & Documents
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
- 316 stainless steel
- Low magnetic permeability
- High corrosion resistance
- High gloss polish
- ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Product Family Options
Weight sets in plastic case, with certificates
Product Information
- RangeNo. of WeightsAvailabilityPrice
- Supplier Number: 11124067
- Supplier Number: 11124081
- Supplier Number: 11124069
- Supplier Number: 11124085
- Supplier Number: 11124071
- Supplier Number: 11124073
- Supplier Number: 11124075
Weight sets in plastic case, without certificates
Product Information
- RangeNo. of WeightsAvailabilityPrice
- Supplier Number: 11124066
- Supplier Number: 11124080
- Supplier Number: 11124068
- Supplier Number: 11124084
- Supplier Number: 11124070
- Supplier Number: 11124072
- Supplier Number: 11124074