xPIPETTE™ Carousel Stand
Carousel stand for xPIPETTE™ pipettes
Catalog # 76320-254
Supplier: Ohaus
Fournisseur #: 30518126
Jump to:
Product Details & Documents
The carousel pipette stand is a rotating carousel stand for the xPIPETTE.
- Fits both single and multichannel models
- Holds seven pipettes total
- Made from chemically resistant polypropylene
Pipettes hang vertically, preventing liquids from contaminating the pipette shaft.
Spécifications
- Désignation:Carousel stand for xPIPETTE™ pipettes