Hanna Instruments pH Maintenance Solutions | Avantor
Hanna Instruments pH Maintenance Solutions
pH maintenance solutions
Catalog # CA470350-702
Supplier:   Hanna
Fournisseur #:  HI54710-11
Hanna Instruments pH Maintenance Solutions
Catalog # CA470350-702
Supplier:  Hanna
CAS Number:  
Hanna Instruments pH Maintenance Solutions
Hanna Instruments pH Maintenance Solutions
pH maintenance solutions
CAS Number:  
Price
$174.75
Log in pour la disponibilité
Pack of 5 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
This is a complete set of all the solutions required for routine pH measurement.
  • 500 ml bottles
  • pH 4.01, 7.01 and 10.01 technical calibration solutions
  • Electrode storage solution and cleaning solution
The set includes pH 4.01, 7.01 and 10.01 technical calibration buffer that are NIST traceable and supplied with a certificate of analysis.
 : pH meter sold separately.
Spécifications
Spécifications
  • Désignation:
    pH maintenance solutions