Eisco® Ring Thrower
Product Details & Documents
Ring thrower kit for studying eddy currents, Lenz Law, and Faraday's Law. Includes two aluminum rings.
- Can be used to demonstrate Eddy currents, Lenz's Law and Faraday's Law
This ring thrower kit is perfect for physics experimentation, including the study of electromagnetism, Eddy Currents, Lenz Law and Faraday's Law. When the aluminum rings are placed on the coil, the changing magnetic field introduces an electric current, which sends the rings into the air. An exciting and interesting experiment for students and teachers or even at home! The ring thrower device measures 12.5" tall, and sits on a base that is 6.25" in diameter.
Delivery information: Set includes two aluminum rings, measuring 2" in diameter and 1" tall.
Specifications
- Désignation:Ring thrower